missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde dimethyl acetal, 97+%

Product Code. 11451567
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451567 100 g 100g
11461567 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451567 Supplier Thermo Scientific Alfa Aesar Supplier No. B24988.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6342-56-9 | C5H10O3 | 118.132 g/mol

Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.

Solubility
Soluble in water. 350 g/L at 20°C.

Notes
Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6342-56-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00008758
InChI Key ULVSHNOGEVXRDR-UHFFFAOYSA-N
Synonym 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
PubChem CID 80650
IUPAC Name 1,1-dimethoxypropan-2-one
SMILES CC(=O)C(OC)OC

Specifications

Melting Point -57°C
Density 0.998
Boiling Point 133°C to 135°C
Flash Point 34°C (93°F)
Refractive Index 1.398
Quantity 100 g
UN Number UN1993
Beilstein 1560557
Solubility Information Soluble in water. 350g/L at 20°C.
Formula Weight 118.13
Percent Purity ≥97%
Chemical Name or Material Pyruvic aldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.