missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde, 35-45% w/w aq. soln

Product Code. 11440528
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440528 500 g 500g
11420528 25 g 25g
11430528 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440528 Supplier Thermo Scientific Alfa Aesar Supplier No. B24664.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78-98-8 | C3H4O2 | 72.06 g/mol

Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

Solubility
Soluble in DMSO, water.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-98-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00006960
InChI Key AIJULSRZWUXGPQ-UHFFFAOYSA-N
Synonym methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
PubChem CID 880
ChEBI CHEBI:17158
IUPAC Name 2-oxopropanal
SMILES CC(=O)C=O

Specifications

Density 1.2
Linear Formula CH3COCHO
Refractive Index 1.405
Quantity 500 g
Beilstein 906750
Sensitivity Air Sensitive
Merck Index 14,6081
Solubility Information Soluble in DMSO,water.
Formula Weight 72.06
Concentration or Composition (by Analyte or Components) 35 to 45% w/w aq. soln
Chemical Name or Material Pyruvic aldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.