Learn More
Pyruvic aldehyde, 35-45% w/w aq. soln
Description
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
Solubility
Soluble in DMSO, water.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Specifications
Specifications
| Density | 1.2 |
| Linear Formula | CH3COCHO |
| Refractive Index | 1.405 |
| Quantity | 25 g |
| Beilstein | 906750 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,6081 |
| Solubility Information | Soluble in DMSO,water. |
| Formula Weight | 72.06 |
| Concentration or Composition (by Analyte or Components) | 35 to 45% w/w aq. soln |
| Show More |
Sicurezza e movimentazione
- Pyruvic aldehyde, 35-45% w/w aqueous solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Pyruvic aldehyde
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.