missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid, TRC

Product Code. 30424954
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424954 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424954 Supplier Toronto Research Chemicals Supplier No. TRCP99889525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyruvic Acid
CAS 127-17-3
Molecular Formula C3 H4 O3
Quantity 25 g
Synonym 2-Oxopropanoic Acid, Pyruvic Acid (8CI), 2-Oxopropanoic acid, 2-Oxopropionic acid, Acetylformic acid, BTS, NSC 179, Pyroracemic acid, α-Ketopropionic acid
InChI Formula InChI=1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)
SMILES CC(=O)C(=O)O
IUPAC Name 2-oxopropanoic acid
Molecular Weight (g/mol) 88.06
Formula Weight 88.016
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.