missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Kinase Antibody (OTI1H4) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30688034 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688034 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688034 Supplier Novus Biologicals Supplier No. NBP344636AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pyruvate Kinase Monoclonal antibody specifically detects Pyruvate Kinase in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyruvate Kinase
Applications Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI1H4
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias PKM, PKM2
Host Species Mouse
Immunogen Full length human recombinant protein of human PKLR (NP_000289) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.