missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Kinase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-44231
This item is not returnable.
View return policy
Description
Specifically detects Pyruvate Kinase in Rabbit samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Pyruvate Kinase Polyclonal specifically detects Pyruvate Kinase in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Pyruvate Kinase | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| PKM, PKM2 | |
| Pyruvate Kinase (Rabbit Muscle) (Uniprot: P11974) | |
| Multi-step | |
| RUO | |
| This product has been reported to react with all forms of pyruvate kinase (pan M-PK). Cross-reactivity against pyruvate kinase from other mammalian tissues is expected but has not been specifically determined. | |
| Rabbit | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:500-1:2000, ELISA 1:4000-1:20000, Immunohistochemistry 1:500-1:2000 | |
| P11974 | |
| Goat | |
| 58.05 kDa | |
| 0.1 mg | |
| Primary | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction