missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30731378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731378 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731378 Supplier Novus Biologicals Supplier No. AF7260JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C Polyclonal antibody specifically detects Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias [Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial, EC 3.1.3, EC 3.1.3.43, MGC119646, PDH, PDP 1, PDPC, PDPC 1, PDPFLJ32517, PPM2CFLJ56179, Protein phosphatase 2C, protein phosphatase 2C, magnesium-dependent, catalytic subunit, pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase, Pyruvate dehydrogenase phosphatase catalytic subunit 1, pyruvate dehyrogenase phosphatase catalytic subunit 1
Host Species Sheep
Immunogen E. coli-derived recombinant human Pyruvate Dehydrogenase Phosphatase/PDP1, Ala72-Glu537, Accession # Q9P0J1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 54704
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.