missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Rabbit anti-Human, Mouse, Rat, Clone: 9A7X4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Pyruvate Dehydrogenase Kinase 1/PDK1 |
|---|---|
| Clone | 9A7X4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18354465
|
Novus Biologicals
NBP3-16720-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380493
|
Novus Biologicals
NBP3-16720-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Monoclonal antibody specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Pyruvate Dehydrogenase Kinase 1/PDK1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5163 | |
| IgG | |
| Affinity purified |
| 9A7X4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
| A synthesized peptide derived from human Pyruvate Dehydrogenase Kinase 1/PDK1/PDHK1 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title