missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen Pyruvate Dehydrogenase Kinase 1/PDK1
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659775
View Documents
Novus Biologicals
NBP2-99975-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18641796
View Documents
Novus Biologicals
NBP2-99975-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal antibody specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human samples. It is validated for ELISA
Specifications

Specifications

Pyruvate Dehydrogenase Kinase 1/PDK1
ELISA
Unconjugated
Rabbit
Autophagy, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction
0.2 um filtered solution in PBS
5163
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
Human
EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial
Produced in rabbits immunized with purified, recombinant Human Pyruvate Dehydrogenase Kinase 1/PDK1 (Accession#: NP_002601.1; Ser29-Ala436)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.