missing translation for 'onlineSavingsMsg'
Learn More

Pyruvate Dehydrogenase E2 Antibody, HRP, Novus Biologicals™

Product Code. 18326735 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326735 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326735 Supplier Novus Biologicals Supplier No. NBP299340H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pyruvate Dehydrogenase E2 Polyclonal specifically detects Pyruvate Dehydrogenase E2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyruvate Dehydrogenase E2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 70 kDa mitochondrial autoantigen of primary biliary cirrhosis, dihydrolipoamide S-acetyltransferase, DLTA, E2 component of pyruvate dehydrogenase complex, EC 2.3.1, M2 antigen complex 70 kDa subunit, PBC, PDC-E2EC 2.3.1.12, PDCE2mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Pyruvate Dehydrogenase E2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1737
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.