missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Dehydrogenase E1 beta subunit Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38354-100ul
This item is not returnable.
View return policy
Description
Pyruvate Dehydrogenase E1 beta subunit Polyclonal antibody specifically detects Pyruvate Dehydrogenase E1 beta subunit in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Pyruvate Dehydrogenase E1 beta subunit | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DKFZp564K0164, mitochondrial, pyruvate dehydrogenase (lipoamide) beta, pyruvate dehydrogenase, E1 beta polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Pyruvate Dehydrogenase E1 beta subunit (NP_000916.2).,, Sequence:, MAAVSGLVRRPLREVSGLLKRRFHWTAPAALQVTVRDAINQGMDEELERDEKVFLLGEEVAQYDGAYKVSRGLWKKYGDKRIIDTPISEMGFAGIAVGAAMAGLRPICEFMTFNFSMQAIDQVINSAAKTYYMSGGLQPVPIVFRGPNGASAGVAAQHSQCFAAWYGHCPGLKVVSPWNSEDAKGLIKSAIRDNNPVVVLENELMYGVPFEFPPEAQSKDFLIPIGKAKIERQGTHITVVSHSRPVGHCLEAAAVLSKEGVECEVINMRTIRPMDMETIEASVMKTNHLVTVEGGWPQFG | |
| 100 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 5162 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction