missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Dehydrogenase E1-alpha subunit Rabbit anti-Human, Mouse, Rat, Clone: 9S3I4, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £423.00
Specifications
| Antigen | Pyruvate Dehydrogenase E1-alpha subunit |
|---|---|
| Clone | 9S3I4 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18347343
|
Novus Biologicals
NBP3-16916-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18326445
|
Novus Biologicals
NBP3-16916-100UL |
100 μg |
£423.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyruvate Dehydrogenase E1-alpha subunit Monoclonal antibody specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| Pyruvate Dehydrogenase E1-alpha subunit | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial | |
| A synthetic phosphorylated peptide around S293 of human Pyruvate Dehydrogenase E1-alpha subunit (P08559). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9S3I4 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Core ESC Like Genes, Lipid and Metabolism, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5160 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title