Learn More
Invitrogen™ Pyruvate Carboxylase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120984
Description
Positive test controls include: HepG2, HeLa, MCF7, A-549, U-251MG, Mouse liver, Mouse brain, Rat liver, Rat brain. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: HSMGLGSKFK EVKKAYVEAN QMLGDLIKVT PSSKIVGDLA QFMVQNGLSR AEAEAQAEEL SFPRSVVEFL QGYIGVPHGG FPEPFRSKVL KDLPRVEGRP GASLPPLDLQ ALEKELVDRH GEEVTPEDVL SAAMYPDVFA HFKDFTATFG PLDSLNTRLF LQGPKIAEEF EVELERGKTL HIKALAVSDL NRAGQRQVFF ELNGQLRSIL VKDTQAMKEM HFHPKALKDV KGQIGAPMPG KVIDIKVVAG AKVAKGQPLC VLSAMKMETV VTSPMEGTVR KVHVTKDMTL EGDDLILEIE.
Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.
Specifications
| Pyruvate Carboxylase | |
| Polyclonal | |
| Unconjugated | |
| PC | |
| EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18517, 18563, 25104, 5091 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11498, P52873, Q05920 | |
| PC, Pcx | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 879-1178 of human Pyruvate carboxylase (Pyruvate carboxylase (PC)) (NP_0010358061). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.