missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvate Carboxylase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £423.00
Specifications
| Antigen | Pyruvate Carboxylase |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000. Use reported in scientific literature (PMID 29937374), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200, KnockDown Validated |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246623
|
Novus Biologicals
NBP1-49536SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18024605
|
Novus Biologicals
NBP1-49536 |
0.1 mL |
£423.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyruvate Carboxylase Polyclonal specifically detects Pyruvate Carboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Pyruvate Carboxylase | |
| Western Blot 1:1000. Use reported in scientific literature (PMID 29937374), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 0.02% Sodium Azide | |
| EC 6.4.1, EC 6.4.1.1, PCBPyruvic carboxylase, pyruvate carboxylase, pyruvate carboxylase, mitochondrial | |
| PC | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen has 96% identity to bovine and porcine. Rat reactivity reported in scientific literature (PMID: 24333689). |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P11498, Q05920 | |
| 5091 | |
| Partial recombinant protein made to an internal region of the human Pyruvate Carboxylase protein (within residues 930-1050). [Swiss-Prot P11498] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 126 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title