missing translation for 'onlineSavingsMsg'
Learn More

Pyrroloquinoline quinone disodium salt, MedChemExpress

Product Code. 16442536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442536

Brand: Medchem Express HY100196A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrroloquinoline quinone disodium salt, a redox co-factor, is an anionic, redox-cycling orthoquinone. Pyrroloquinoline quinone disodium salt is isolated from cultures of methylotropic bacteria and tissues of mammals. Pyrroloquinoline quinone disodium salt is an essential nutrient for mammals and is important for immune function.

Pyrroloquinoline quinone disodium salt, a redox co-factor, is an anionic, redox-cycling orthoquinone. Pyrroloquinoline quinone disodium salt is isolated from cultures of methylotropic bacteria and tissues of mammals. Pyrroloquinoline quinone disodium salt is an essential nutrient for mammals and is important for immune function.

Specifications

Chemical Name or Material Pyrroloquinoline quinone disodium salt
CAS 122628-50-6
Color Red
Purity Grade Notes Research
Molecular Formula C14H4N2Na2O8
Quantity 25 mg
Synonym PQQ disodium saltMethoxatin disodium salt
Solubility Information H2O : 1.25 mg/mL (3.34 mM; Need ultrasonic)
SMILES O=C(C(N1)=CC(C2=O)=C1C3=C(N=C(C(O[Na])=O)C=C3C(O)=O)C2=O)O[Na]
Molecular Weight (g/mol) 374.17
Formula Weight 374.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.