missing translation for 'onlineSavingsMsg'
Learn More

Pyrroloquinoline quinone disodium salt, MedChemExpress

Product Code. 16495577
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495577 5 mg 5mg
16474246 100 mg 100mg
16452838 10 mg 10mg
16442536 25 mg 25mg
16419496 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495577 Supplier Medchem Express Supplier No. HY100196A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrroloquinoline quinone disodium salt, a redox co-factor, is an anionic, redox-cycling orthoquinone. Pyrroloquinoline quinone disodium salt is isolated from cultures of methylotropic bacteria and tissues of mammals. Pyrroloquinoline quinone disodium salt is an essential nutrient for mammals and is important for immune function.

Pyrroloquinoline quinone disodium salt, a redox co-factor, is an anionic, redox-cycling orthoquinone. Pyrroloquinoline quinone disodium salt is isolated from cultures of methylotropic bacteria and tissues of mammals. Pyrroloquinoline quinone disodium salt is an essential nutrient for mammals and is important for immune function.

Specifications

Chemical Name or Material Pyrroloquinoline quinone disodium salt
CAS 122628-50-6
Color Red
Purity Grade Notes Research
Molecular Formula C14H4N2Na2O8
Quantity 5 mg
Synonym PQQ disodium saltMethoxatin disodium salt
Solubility Information H2O : 1.25 mg/mL (3.34 mM; Need ultrasonic)
SMILES O=C(C(N1)=CC(C2=O)=C1C3=C(N=C(C(O[Na])=O)C=C3C(O)=O)C2=O)O[Na]
Molecular Weight (g/mol) 374.17
Formula Weight 374.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.