missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyrroloquinoline quinone

Product Code. 17211292 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17211292 250 mg 250mg
17201292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17211292 Supplier Thermo Scientific Chemicals Supplier No. 467012500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72909-34-3 | C14H6N2O8, C14H6N2O8 | 330.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72909-34-3
Molecular Formula C14H6N2O8

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: slightly soluble.
Formula Weight 330.21
InChI Key MMXZSJMASHPLLR-UHFFFAOYSA-N
SMILES OC(=O)C1=CC2=C(N1)C1=C(C=C(N=C1C(=O)C2=O)C(O)=O)C(O)=O
IUPAC Name 4,5-dioxo-1H,4H,5H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Molecular Weight (g/mol) 330.21
Chemical Name or Material Pyrroloquinoline quinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.