missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidinedithiocarbamate ammonium, MedChemExpress

Product Code. 16477526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477526 Supplier Medchem Express Supplier No. HY18738
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyrrolidinedithiocarbamate ammonium (Ammonium pyrrolidinedithiocarbamate) is a selective and blood-brain barrier (BBB) permeable NF-κB inhibitor.

Pyrrolidinedithiocarbamate ammonium (Ammonium pyrrolidinedithiocarbamate) is a selective and blood-brain barrier (BBB) permeable NF-κB inhibitor.

Specifications

Chemical Name or Material Pyrrolidinedithiocarbamate ammonium
Physical Form Solid
CAS 5108-96-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H12N2S2
For Use With (Application) Cancer-programmed cell death
Synonym Ammonium pyrrolidinedithiocarbamatePDTC ammonium APDC
Solubility Information H2O : 50 mg/mL (304.34 mM; Need ultrasonic)
SMILES SC(N1CCCC1)=S.N
Molecular Weight (g/mol) 164.29
Formula Weight 164.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.