missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99+%, Thermo Scientific™

Click to view available options
Quantity:
5mL
100mL
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
123-75-1
C4H10ClN
107.58
MFCD00005249
FCLZCOCSZQNREK-UHFFFAOYSA-N
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
31268
CHEBI:33135
pyrrolidine
[H+].[Cl-].C1CCNC1
This item is not returnable. View return policy
123-75-1
C4H10ClN
107.58
MFCD00005249
FCLZCOCSZQNREK-UHFFFAOYSA-N
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
31268
CHEBI:33135
pyrrolidine
[H+].[Cl-].C1CCNC1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 0.8660g/mL
Melting Point -63.0°C
Boiling Point 86.0°C to 88.0°C
Flash Point 3°C
Assay Percent Range 99% min. (GC)
Beilstein 20,4
Packaging Plastic drum
Fieser 01,972; 02,354; 03,240; 07,309; 08,428; 13,14
Merck Index 15,8129
Refractive Index 1.4421 to 1.4441
Quantity 10L
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
Specific Gravity 0.866
Formula Weight 71.11
Percent Purity 99+%
Viscosity 0.94 mPa.s (20°C)
Chemical Name or Material Pyrrolidine, 99+%
Show More Show Less

RUO – Research Use Only