missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99+%, Thermo Scientific Chemicals

Catalog Number 10120312
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
123-75-1
C4H10ClN
107.58
MFCD00005249
FCLZCOCSZQNREK-UHFFFAOYSA-N
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
31268
CHEBI:33135
[H+].[Cl-].C1CCNC1
This item is not returnable. View return policy
123-75-1
C4H10ClN
107.58
MFCD00005249
FCLZCOCSZQNREK-UHFFFAOYSA-N
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
31268
CHEBI:33135
[H+].[Cl-].C1CCNC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8660g/mL
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 mL
Fieser 01,972; 02,354; 03,240; 07,309; 08,428; 13,14
IUPAC Name hydrogen pyrrolidine chloride
Formula Weight 71.11
Chemical Name or Material Pyrrolidine, 99+%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'