missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-3-carboxylic acid, TRC

Product Code. 30425914
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425914 10 g 10g
30424097 5 g 5g
30417339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425914 Supplier Toronto Research Chemicals Supplier No. TRCP99994810G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59378-87-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
IUPAC Name pyrrolidine-3-carboxylic acid
SMILES OC(=O)C1CCNC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H9NO2/c7-5(8)4-1-2-6-3-4/h4,6H,1-3H2,(H,7,8)
Formula Weight 115.0633
Chemical Name or Material Pyrrolidine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.