missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-3-carboxylic acid, 98%

Product Code. 15559473 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15559473 5 g 5g
15549473 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15559473 Supplier Thermo Scientific Acros Supplier No. 459110050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59378-87-9 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59378-87-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
InChI Key JAEIBKXSIXOLOL-UHFFFAOYSA-N
Synonym 3-pyrrolidinecarboxylic acid, 3-pyrrolidine carboxylic acid, beta-proline, 3-pyrrolidinecarboxylicacid, pyrrolidine-3-carboxylicacid, dl-pyrrolidine-3-carboxylic acid, beta-dl-proline, pubchem10439, acmc-209mcv, acmc-20a9vh
PubChem CID 3034645
IUPAC Name pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 182.0°C to 184.0°C
Elemental Analysis 50.86 to max. 53.47% min. Carbon 11.86 to max. 12.47% min. Nitrogen
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material Pyrrolidine-3-carboxylic acid
compliance-icons
Product Identifier
  • Pyrrolidine-3-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.