missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxylic acid, MedChemExpress

Product Code. 16413419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16413419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16413419 Supplier Medchem Express Supplier No. HYW001963
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Pyrrole-2-carboxylic acid is a natural alkaloid from the marine bacterium Pelomonas puraquae sp. Nov.

Pyrrole-2-carboxylic acid is a natural alkaloid from the marine bacterium Pelomonas puraquae sp. Nov.

Specifications

Chemical Name or Material Pyrrole-2-carboxylic acid
CAS 634-97-9
Color Gray
Purity Grade Notes Research
Molecular Formula C5H5NO2
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (900.09 mM) ∣H2O : 5.88 mg/mL (52.93 mM; Need ultrasonic)
SMILES OC(C1=CC=CN1)=O
Molecular Weight (g/mol) 111.1
Formula Weight 111.1
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.