missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxylic acid, 97%

Product Code. 10239133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10239133 1 g Glass bottle 1g
10590224 5 g Glass bottle 5g
10107673 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239133 Supplier Thermo Scientific Acros Supplier No. 132070010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 634-97-9 | C5H5NO2 | 111.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-97-9
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00005219
InChI Key WRHZVMBBRYBTKZ-UHFFFAOYSA-N
Synonym pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc
PubChem CID 12473
ChEBI CHEBI:36751
IUPAC Name 1H-pyrrole-2-carboxylic acid
SMILES C1=CNC(=C1)C(=O)O

Specifications

Melting Point 207.0°C to 208.0°C
Color Pink to White
Density 0.862g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4410 to 1.4450
Quantity 1 g
Beilstein 22, 22
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether
Formula Weight 111.1
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Pyrrole-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Pyrrole-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.