missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxaldehyde, 99%

Product Code. 11455233
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455233 10 g 10g
11453990 25 g 25g
11463990 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455233 Supplier Thermo Scientific Alfa Aesar Supplier No. A12838.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pyrrole-2-carboxaldehyde, 99%, C5H5NO, CAS Number-1003-29-8, 2-pyrrolecarboxaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolaldehyde, 1h-pyrrole-2-carboxaldehyde, alpha-pyrrolaldehyde, 2-formylpyrrole, 2-pyrrolylcarboxaldehyde, pyrrole-2-aldehyde, pyrrole-2-carboxaldehyde, pyrrole-2-carbaldehyde, 100g | CAS: 1003-29-8 | C5H5NO | 95.101 g/mol

Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-29-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00005217
InChI Key ZSKGQVFRTSEPJT-UHFFFAOYSA-N
Synonym pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
PubChem CID 13854
ChEBI CHEBI:59978
IUPAC Name 1H-pyrrole-2-carbaldehyde
SMILES C1=CNC(=C1)C=O

Specifications

Melting Point 40°C to 46°C
Boiling Point 217°C to 219°C
Flash Point 106°C (222°F)
Quantity 10 g
Beilstein 105745
Sensitivity Air sensitive
Solubility Information Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 95.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.