missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYROXD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584757
This item is not returnable.
View return policy
Description
Immunogen sequence: VLTAPITKVL DQWFESEPLK ATLATDAVIG AMTSPHTPGS GYVLLHHVMG GLEGMQGAWG YVQGGMGALS DAIASSATTH GASIFTE Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 95%.
PYROXD2 is a protein coding gene.
Specifications
| PYROXD2 | |
| Polyclonal | |
| Unconjugated | |
| PYROXD2 | |
| 3830409H07Rik; 4833409A17Rik; C10orf33; FP3420; Phytn_dehydro and Pyr_redox domain containing protein RGD1303232; pyridine nucleotide-disulfide oxidoreductase domain-containing protein 2; pyridine nucleotide-disulphide oxidoreductase domain 2; Pyroxd2; RGD1303232 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N2H3 | |
| PYROXD2 | |
| Recombinant protein corresponding to Human PYROXD2. Recombinant protein control fragment (Product #RP-105424). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction