missing translation for 'onlineSavingsMsg'
Learn More

Pyrophosphoryl chloride, Thermo Scientific Chemicals

Product Code. 11358027
Click to view available options
Quantity:
0.5 g
5 g
25 g
Unit Size:
0.5g
25g
5g
This item is not returnable. View return policy

Product Code. 11358027

Brand: Thermo Scientific Alfa Aesar 022362.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pyrophosphoryl chloride serves as a reductive chlorination reagent used in the synthesis of quetiapine. It acts as a phosphorylating agent involved in nucleoside synthesis. It plays an important role in Vilsmeier formylation and glyoxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13498-14-1
Molecular Formula Cl4O3P2
Molecular Weight (g/mol) 251.745
MDL Number MFCD00061542
InChI Key CNTIXUGILVWVHR-UHFFFAOYSA-N
Synonym diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
PubChem CID 4432411
SMILES O=P(OP(=O)(Cl)Cl)(Cl)Cl

Specifications

Boiling Point 101°C (10mmHg)
Melting Point -17°C
Color Colorless through Yellow
Physical Form Liquid
Quantity 0.5 g
UN Number UN3264
Solubility Information Reacts violently with waterMiscible with most organic solvents.
Formula Weight 251.76
Odor Acrid
Sensitivity Air & Moisture Sensitive
Density 1.82g/mL
Chemical Name or Material Pyrophosphoryl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.