missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyronin Y, pure, certified

Product Code. 10138090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10138090 5 g Glass bottle 5g
10128090 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10138090 Supplier Thermo Scientific Chemicals Supplier No. 418630050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-32-0 | C17H19ClN2O | 302.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-32-0
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00011725
InChI Key INCIMLINXXICKS-UHFFFAOYSA-M
Synonym C.I. 45005, Pyronin G
PubChem CID 7085
ChEBI CHEBI:87347
IUPAC Name [6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]

Specifications

Melting Point 250.0°C to 260.0°C
Absorption Ratio 1.23 to 1.7
Dye Content 45% min.
Beilstein 18, V,10, 181
Merck Index 15, 8119
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Lambda Max 546 to 551nm
Formula Weight 302.79
Grade Pure
Packaging Glass bottle
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Pyronin Y
Show More Show Less
compliance-icons
Product Identifier
  • Pyronin Y
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.