missing translation for 'onlineSavingsMsg'
Learn More

Pyronin B, 50% dye content, Thermo Scientific™

Product Code. 15535876 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535876 25 g 25g
15525876 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535876 Supplier Thermo Scientific Acros Supplier No. 191390250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1229438-47-4
Molecular Formula C21H27Cl4FeN2O
Molecular Weight (g/mol) 521.11

Specifications

Melting Point 174.0°C to 176.0°C
Quantity 25 g
Beilstein 18,V,10,182
Merck Index 15,8118
Formula Weight 521.11
Chemical Name or Material Pyronin B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.