missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic acid, 96%

Product Code. 11496296
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496296 100 g 100g
11406306 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496296 Supplier Thermo Scientific Alfa Aesar Supplier No. B23099.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-05-4 | C10H6O8 | 254.15 g/mol

Pyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.

Solubility
Soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-05-4
Molecular Formula C10H6O8
Molecular Weight (g/mol) 254.15
MDL Number MFCD00002471
InChI Key CYIDZMCFTVVTJO-UHFFFAOYSA-N
Synonym pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
PubChem CID 6961
ChEBI CHEBI:45165
IUPAC Name benzene-1,2,4,5-tetracarboxylic acid
SMILES C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 281°C to 285°C
Quantity 100 g
Merck Index 14,8004
Solubility Information Soluble in alcohol.
Formula Weight 254.15
Percent Purity 96%
Chemical Name or Material Pyromellitic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.