missing translation for 'onlineSavingsMsg'
Learn More

Pyromeconic Acid, TRC

Product Code. 30415971
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415971 100 mg 100mg
30413897 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415971 Supplier Toronto Research Chemicals Supplier No. TRCP997600100MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-63-9
Molecular Formula C5 H4 O3
Molecular Weight (g/mol) 112.08
Synonym 3-Hydroxy-4H-pyran-4-one, 3-Hydroxy-4-pyrone, 3-Hydroxypyran-4-one, NSC 78608, Pyrocomenic Acid
IUPAC Name 3-hydroxypyran-4-one
SMILES OC1=COC=CC1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H4O3/c6-4-1-2-8-3-5(4)7/h1-3,7H
Formula Weight 112.016
Chemical Name or Material Pyromeconic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.