missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, ACS reagent

Product Code. 10195173 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195173 2.5 kg Plastic bottle 2.5kg
10391111 100 g Plastic bottle 100g
10301121 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10195173 Supplier Thermo Scientific Acros Supplier No. 418620025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131.0°C to 135.0°C
Color White
Boiling Point 309.0°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula C6H3(OH)3
Quantity 2.5 kg
UN Number 2811
Beilstein 06, 1071
Merck Index 15, 8112
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide
Formula Weight 126.11
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Pyrogallol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyrogallol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.