missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, MedChemExpress

Product Code. 16402579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402579 Supplier Medchem Express Supplier No. HYN1579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrogallol is a polyphenol compound, which has anti-fungal and anti-psoriatic properties. Pyrogallol is a reductant that is able to generate free radicals, in particular superoxide anions.

Pyrogallol is a polyphenol compound, which has anti-fungal and anti-psoriatic properties. Pyrogallol is a reductant that is able to generate free radicals, in particular superoxide anions.

Specifications

Chemical Name or Material Pyrogallol
Physical Form Solid
CAS 87-66-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H6O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (792.96 mM) ∣H2O : 50 mg/mL (396.48 mM; Need ultrasonic)
SMILES OC1=CC=CC(O)=C1O
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.