missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, Extra Pure, SLR, Fisher Chemical

Product Code. 10306340 Shop All Fisher Chemical Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Packaging:
Glass Jar
missing translation for 'unitSize':
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
10306340 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10306340 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' P/8120/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 87-66-1
Melting Point 131°C
Color White
pH 5.8
Boiling Point 309°C
Packaging Glass Jar
Molecular Formula C6H6O3
MDL Number MFCD00002192
Quantity 100 g
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILES OC1=CC=CC(O)=C1O
IUPAC Name benzene-1,2,3-triol
Molecular Weight (g/mol) 126.11
PubChem CID 1057
ChEBI CHEBI:16164
Formula Weight 126.11g/mol
Vapor Pressure 13.3 hPa 168
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyrogallol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.