missing translation for 'onlineSavingsMsg'
Learn More

Pyrithione, MedChemExpress

Product Code. 16458806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458806 Supplier Medchem Express Supplier No. HYB1747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrithione, a Transition metal complexe, is a zinc ionophore that causes increased zinc levels within mammalian cells. Pyrithione has potent bactericidal and anti-fungal activity.

Pyrithione, a Transition metal complexe, is a zinc ionophore that causes increased zinc levels within mammalian cells. Pyrithione has potent bactericidal and anti-fungal activity.

Specifications

Chemical Name or Material Pyrithione
Physical Form Solid
CAS 1121-30-8
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H5NOS
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (983.01 mM; Need ultrasonic)
SMILES S=C1C=CC=CN1O
Molecular Weight (g/mol) 127.16
Formula Weight 127.16
Percent Purity 96.99%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.