Learn More
Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in)
Description
For grain mounts, XRD and microprobe standards, identification of unknown minerals.Pyrite is used in the preparation of iron(II) sulfate, sulfur dioxide and iron(II) sulfide. It finds application as semiconductor material, as the cathode material in energizer band and non-rechargeable lithium batteries. It is utilized to make marcasite jewelry. In addition, it is used in photovoltaic solar panels. It is useful for grain mounts, XRD and microprobe standards and identification of unknown minerals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Grains or Chunks |
| Quantity | 25 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 119.98 |
| Refractive Index | 3.08 |
| Appearance | Grains/chunks |
| Density | 5.02 g/cm3 at 20°C |
| Chemical Name or Material | Pyrite |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.