missing translation for 'onlineSavingsMsg'
Learn More

Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in)

Product Code. 11309258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309258 1 g 1g
11399248 5 g 5g
11319258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309258 Supplier Thermo Scientific Alfa Aesar Supplier No. 042633.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals | CAS: 1309-36-0 | FeS2 | 119.965 g/mol

For grain mounts, XRD and microprobe standards, identification of unknown minerals.Pyrite is used in the preparation of iron(II) sulfate, sulfur dioxide and iron(II) sulfide. It finds application as semiconductor material, as the cathode material in energizer band and non-rechargeable lithium batteries. It is utilized to make marcasite jewelry. In addition, it is used in photovoltaic solar panels. It is useful for grain mounts, XRD and microprobe standards and identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Naturally occurring mineral; Hardness: 6-6.5
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-36-0
Molecular Formula FeS2
Molecular Weight (g/mol) 119.965
MDL Number MFCD00064690
InChI Key NFMAZVUSKIJEIH-UHFFFAOYSA-N
Synonym iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
PubChem CID 14788
IUPAC Name bis(sulfanylidene)iron
SMILES S=[Fe]=S

Specifications

Physical Form Grains or Chunks
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 119.98
Refractive Index 3.08
Appearance Grains/chunks
Density 5.02
Chemical Name or Material Pyrite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.