missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine, 99%

Product Code. 10656354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656354 25 g Glass bottle 25g
10001840 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656354 Supplier Thermo Scientific Acros Supplier No. 157700250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289-95-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006059
InChI Key CZPWVGJYEJSRLH-UHFFFAOYSA-N
Synonym 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
PubChem CID 9260
ChEBI CHEBI:16898
IUPAC Name pyrimidine
SMILES C1=CN=CN=C1

Specifications

Melting Point 20°C to 22°C
Color Colorless to Orange
Density 1.0100g/mL
Boiling Point 123°C to 124°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5010 to 1.504
Quantity 25 g
Beilstein 23, 89
Merck Index 15, 8100
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 80.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • Pyrimidine
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.