missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine, 98+%

Product Code. 11482957
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482957 1 g 1g
11492957 5 g 5g
11473990 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482957 Supplier Thermo Scientific Alfa Aesar Supplier No. A15534.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol

Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289-95-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006059
InChI Key CZPWVGJYEJSRLH-UHFFFAOYSA-N
Synonym 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
PubChem CID 9260
ChEBI CHEBI:16898
IUPAC Name pyrimidine
SMILES C1=CN=CN=C1

Specifications

Melting Point 20°C to 22°C
Density 1.016
Boiling Point 123°C to 124°C
Flash Point 31°C (87°F)
Refractive Index 1.504
Quantity 1 g
UN Number UN1993
Beilstein 103894
Sensitivity Hygroscopic
Merck Index 14,7987
Solubility Information Fully miscible in water.
Formula Weight 80.09
Percent Purity 99%
Chemical Name or Material Pyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.