missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid hemihydrate, 97%, Thermo Scientific Chemicals

£68.10 - £221.00

Chemical Identifiers

CAS 696602-91-2
Molecular Formula C8H12B2N4O5
Molecular Weight (g/mol) 265.827
MDL Number MFCD03002366
InChI Key YXMKGFBYXBTOJS-UHFFFAOYSA-N
Synonym pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
PubChem CID 67112167
IUPAC Name pyrimidin-5-ylboronic acid;hydrate
SMILES B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482260
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H29212.03
1 g
£68.10
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11492260
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H29212.09
10 g
£221.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

696602-91-2
265.827
YXMKGFBYXBTOJS-UHFFFAOYSA-N
67112167
B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
C8H12B2N4O5
MFCD03002366
pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
pyrimidin-5-ylboronic acid;hydrate

Specifications

696602-91-2
C8H12B2N4O5
pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O
265.827
132.91 (123.91 Anhydrous)
Pyrimidine-5-boronic acid hemihydrate
>300°C
MFCD03002366
YXMKGFBYXBTOJS-UHFFFAOYSA-N
pyrimidin-5-ylboronic acid;hydrate
67112167
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only