missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid hemihydrate, 97%, Thermo Scientific Chemicals

Product Code. 11492260
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696602-91-2
Molecular Formula C8H12B2N4O5
Molecular Weight (g/mol) 265.827
MDL Number MFCD03002366
InChI Key YXMKGFBYXBTOJS-UHFFFAOYSA-N
Synonym pyrimidin-5-ylboronic acid hemihydrate, pyrimidine-5-boronic acid hemihydrate, 132.91 anhydrous, c4h5bn2o2.5h2o, bis pyrimidin-5-ylboronic acid hydrate
PubChem CID 67112167
IUPAC Name pyrimidin-5-ylboronic acid;hydrate
SMILES B(C1=CN=CN=C1)(O)O.B(C1=CN=CN=C1)(O)O.O

Specifications

Melting Point >300°C
Quantity 10 g
Formula Weight 132.91 (123.91 Anhydrous)
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-boronic acid hemihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'