missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-thiocarboxamide, 97%

Product Code. 15470978
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470978 1 g 1g
15490978 250 mg 250mg
15480978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470978 Supplier Thermo Scientific Alfa Aesar Supplier No. H54783.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4537-73-9 | C5H5N3S | 139.176 g/mol

Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4537-73-9
Molecular Formula C5H5N3S
Molecular Weight (g/mol) 139.176
MDL Number MFCD12093703
InChI Key QAUHVPUYFSGVME-UHFFFAOYSA-N
Synonym 2-pyrimidinecarbothioamide, pyrimidine-2-thiocarboxamide, 2-pyrimidinecarbothioamide 9ci
PubChem CID 23273397
IUPAC Name pyrimidine-2-carbothioamide
SMILES C1=CN=C(N=C1)C(=S)N

Specifications

Quantity 1 g
Beilstein 508162
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material Pyrimidine-2-thiocarboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.