missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-2-carboxylic acid, 95%

Product Code. 15730899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730899 5 g 5g
15720899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15730899 Supplier Thermo Scientific Acros Supplier No. 455830050
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31519-62-7
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856161
InChI Key ZFCHNZDUMIOWFV-UHFFFAOYSA-N
Synonym 2-pyrimidinecarboxylic acid, 2-carboxypyrimidine, pyrimidine carboxylic acid, pyrimidinecarboxylic acid, 2-carboxy-1,3-diazine, 2-pyrimidinecarboxylic acid 6ci,8ci,9ci, 2-pyrimidinecarboxylicacid, 2-carboxy pyrimidine, pubchem9603, pyrimidine-carboxylic acid
PubChem CID 12626245
IUPAC Name pyrimidine-2-carboxylic acid
SMILES OC(=O)C1=NC=CC=N1

Specifications

Melting Point 190.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 124.1
Percent Purity 95%
Chemical Name or Material Pyrimidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.