missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pyrimethamine

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Potent inhibitor of multidrug and toxin extrusion (MATE) transporters (Ki values are 46 and 77 nM for human MATE2-K-HEK293 and MATE1-HEK293 cells respectively). Also inhibits dihydrofolate reductase (DHFR) and STAT3. Decreases proliferation of human autosomal dominant polycystic kidney disease cells.
Specifications
Specifications
| CAS | 58-14-0 |
| Quantity | 50 mg |
| Molecular Formula | C12H13ClN4 |
| Formula Weight | 248.71 |
| Purity | >99% |
| Molecular Weight (g/mol) | 248.71 |
| Chemical Name or Material | Pyrimethamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction