missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrimethamine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pyrimethamine
Specifications
Specifications
| Chemical Name or Material | Pyrimethamine |
| CAS | 58-14-0 |
| Analyte or Component Names | Pyrimethamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H13 Cl N4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Pyrimethamine, 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 2,4-Diamino-5-(4-chlorophenyl)-6-ethylpyrimidine, 2,4-Diamino-5-p-chlorophenyl-6-ethylpyrimidine, 4753 R.P., 5-(4-Chlorophenyl)-2,4-diamino-6-ethylpyrimidine, 5-(P-Chlorophenyl)-6-ethyl-2,4-diaminopyrimidine, BW 50-63, Chloridin, Chloridine, Darachlor, Darapram, Daraprim, Daraprime, Diaminopyritamin, Erbaprelina, Khloridin, Malacid, Malocid, Malocide, NSC 3061, Pirimecidan, Pirimetamin, Pyrimethamin, Pyrimethamine, RP 4753, Tindurin, Tinduring, WR 2978 |
| InChI Formula | InChI=1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17) |
| SMILES | CCc1nc(N)nc(N)c1c2ccc(Cl)cc2 |
| IUPAC Name | 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction