missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, MedChemExpress

Product Code. p-200096345
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16447426

Brand: Medchem Express HY18062

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrimethamine(RP4753) is a medication used for protozoal infections; interferes with tetrahydrofolic acid synthesis from folic acid by inhibiting the enzyme dihydrofolate reductase (DHFR).

Pyrimethamine(RP4753) is a medication used for protozoal infections

Specifications

Chemical Name or Material Pyrimethamine
CAS 58-14-0
Color White
Purity Grade Notes Research
Molecular Formula C12H13ClN4
Quantity 100 mg
Synonym Pirimecidan Pirimetamin RP 4753
Solubility Information DMSO : 20 mg/mL (80.41 mM; Need ultrasonic)
SMILES NC1=NC(N)=C(C2=CC=C(Cl)C=C2)C(CC)=N1
Molecular Weight (g/mol) 248.71
Formula Weight 248.71
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.