missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, LoGiCal

Product Code. 30460624
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30460624

Brand: LoGiCal LGCFOR1630.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrimethamine
CAS 58-14-0
Analyte or Component Names Pyrimethamine
Shipping Condition Room Temperature
Molecular Formula C12 H13 Cl N4
Certifications/Compliance ISO 9001
Synonym Pyrimethamine, 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 2,4-Diamino-5-(4-chlorophenyl)-6-ethylpyrimidine, 2,4-Diamino-5-p-chlorophenyl-6-ethylpyrimidine, 4753 R.P., 5-(4-Chlorophenyl)-2,4-diamino-6-ethylpyrimidine, 5-(P-Chlorophenyl)-6-ethyl-2,4-diaminopyrimidine, BW 50-63, Chloridin, Chloridine, Darachlor, Darapram, Daraprim, Daraprime, Diaminopyritamin, Erbaprelina, Khloridin, Malacid, Malocid, Malocide, NSC 3061, Pirimecidan, Pirimetamin, Pyrimethamin, Pyrimethamine, RP 4753, Tindurin, Tinduring, WR 2978
InChI Formula InChI=1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)
Solution Type Neat
SMILES CCc1nc(N)nc(N)c1c2ccc(Cl)cc2
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 248.71
Quantity 10 mg
Formula Weight 248.0829 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.