missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, 100.5%, MP Biomedicals™

Product Code. 11450742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450742 25 g 25g
11430742 1 g 1g
11440742 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450742 Supplier MP Biomedicals Supplier No. 0219418025
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4
Molecular Weight (g/mol) 248.71
MDL Number MFCD00057350
InChI Key WKSAUQYGYAYLPV-UHFFFAOYSA-N
Synonym pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
PubChem CID 4993
ChEBI CHEBI:8673
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

Specifications

Color White
Assay Percent Range 1.005
Quantity 25 g
Formula Weight 248.7
Physical Form Powder
Chemical Name or Material Pyrimethamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.