missing translation for 'onlineSavingsMsg'
Learn More

Pyrilamine maleate

Product Code. 15415519
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415519 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415519 Supplier Thermo Scientific Alfa Aesar Supplier No. J63247.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective inverse agonist for the H-1 histamine receptor | CAS: 59-33-6 | C21H27N3O5 | 401.46 g/mol

Pyrilamine maleate Selective inverse agonist for the H-1 histamine receptor. Pyrilamine Maleate is commonly utilized as a radioligand ([3H]Mepyramine) binding assay for the H1 receptor. And also used to exhibit blocking ability of KCNQ/M channels which is potentially related to an adverse effect seen in excess intake of antihistamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrilamine maleate Selective inverse agonist for the H-1 histamine receptor. Pyrilamine Maleate is commonly utilized as a radioligand ([3H]Mepyramine) binding assay for the H1 receptor. And also used to exhibit blocking ability of KCNQ/M channels which is potentially related to an adverse effect seen in excess intake of antihistamines.

Solubility
Soluble in water at 100mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-33-6
Color White
Molecular Formula C21H27N3O5
MDL Number MFCD00069333
Quantity 25 g
Merck Index 14,7984
Synonym pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
Solubility Information Soluble in water at 100mg/ml
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
SMILES OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1
IUPAC Name (2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Molecular Weight (g/mol) 401.46
PubChem CID 5284451
Formula Weight 401.46
Physical Form Powder
Chemical Name or Material Pyrilamine maleate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.