missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxylamine, MedChemExpress

Product Code. 16448806
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16448806

Brand: Medchem Express HYB1745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyridoxylamine is an advanced glycation end production (AGEs) and lipoxidation end products (ALEs) inhibitor, to protect against diabetes-induced retinal vascular lesions.

Pyridoxylamine is an advanced glycation end production (AGEs) and lipoxidation end products (ALEs) inhibitor, to protect against diabetes-induced retinal vascular lesions.

Specifications

Chemical Name or Material Pyridoxylamine
CAS 85-87-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H12N2O2
Quantity 100 mg
Solubility Information DMSO : 16.67 mg/mL (99.11 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (37.16 mM; Need ultrasonic)
SMILES OCC1=C(CN)C(O)=C(C)N=C1
Molecular Weight (g/mol) 168.19
Formula Weight 168.19
Percent Purity 99.14%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.